EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Reactive silver inks for patterning high-conductivity features at mild temperatures.
PMID 22220580
Abstract
Reactive silver inks for printing highly conductive features (>10(4) S/cm) at room temperature have been created. These inks are stable, particle-free, and suitable for a wide range of patterning techniques. Upon annealing at 90 °C, the printed electrodes exhibit an electrical conductivity equivalent to that of bulk silver.