EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacy and pharmacology
Comparison of sildenafil with strontium fructose diphosphate in improving erectile dysfunction against upregulated cavernosal NADPH oxidase, protein kinase Cε, and endothelin system in diabetic rats.
PMID 22221100
Abstract
Phosphodiesterase type 5 inhibitors are potent in relieving erectile dysfunction (ED), however, they are less satisfactory in diabetic patients, probably due to the pro-inflammatory biomarkers caused by diabetes. Therefore, it was interesting to compare the effects of sildenafil with strontium fructose 1,6-diphosphate (FDP-Sr) on cavernosal vascular activity and expressions of pro-inflammatory biomarkers in diabetic rats. Male Sprague-Dawley rats were injected with streptozocin (60 mg/kg, i.p.) to develop diabetes. The animals were diabetic for eight weeks with sildenafil (12 mg/kg per day) or FDP-Sr (200 mg/kg per day) being administered for the last four of those eight weeks. Sildenafil was more effective in relieving reduced vascular dilatation (relevant to ED), but less in attenuating over-expressions of NADPH oxidase p22, p47 and p67 subunits, and ET(A/B) R (endothelin receptor type A and type B) in the diabetic cavernosum. In contrast, FDP-Sr was less effective in improving ED, but more effective in normalizing the abnormal NADPH oxidase and ET(A/B) R. The activated NADPH oxidase and upregulated ET(A) R and ET(B) R due to diabetic lesions played a minor or moderate role in ED. By offering extra ATP, FPD-Sr suppressed these abnormalities, however, sildenafil did not. A combined therapy of sildenafil with FDP-Sr may be more effective in relieving ED in diabetic patients through normalizing pro-inflammatory cytokines and improving the nitric oxide/cGMP pathway in the cavernosum.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6803
D-Fructose 1,6-bisphosphate trisodium salt hydrate, ≥98% (TLC)
C6H11Na3O12P2 · xH2O