EMAIL THIS PAGE TO A FRIEND
Helicobacter
A comparison between sequential therapy and a modified bismuth-based quadruple therapy for Helicobacter pylori eradication in Iran: a randomized clinical trial.
PMID 22221615
Abstract
Sequential regimens have been recently reported to be superior to the standard triple therapies in Helicobacter pylori eradication, but most of these studies were performed in Europe and data from developing countries are lacking. So we designed a study to compare a sequential regimen with a bismuth-based quadruple therapy that contains a short course of furazolidone, in Iran. Two hundred and ninety-six patients with duodenal ulcer and naïve H. pylori infection were randomized into two groups: 148 patients received (PAB-F) pantoprazole (40mg-bid), amoxicillin (1g-bid), and bismuth subcitrate (240mg-bid) for 2weeks and furazolidone (200mg-bid) just during the first week. And 148 patients received (PA-CT) pantoprazole (40mg-bid) for 10days, amoxicillin (1g-bid) for the first 5days, and clarithromycin (500mg-bid) plus tinidazole (500mg-bid) just during the second 5days. C(14) -urea breath test was performed 8weeks after the treatment. Two hundred and sixty-one patients completed the study (137 patients in the PA-CT and 124 in the PAB-F group). The results were not statistically different between the two groups in the eradication rates and the severity of side effects. The intention to treat eradication rate was 80.4% in the PAB-F group and 83.7% in the PA-CT group. Per-protocol eradication rates were 88.7% and 89.1%, respectively. Because the two regimens showed acceptable and similar abilities in H. pylori eradication and because of much higher cost of clarithromycin in Iran, the furazolidone containing regimen seems to be superior. Further modifications of sequential therapies are needed to make them ideal regimens in developing countries.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9505
Furazolidone
C8H7N3O5
46297
Furazolidone, VETRANAL, analytical standard
C8H7N3O5