EMAIL THIS PAGE TO A FRIEND
Revista espanola de medicina nuclear e imagen molecular
[Translational molecular imaging: an investment for the future].
PMID 22222021
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
158186
Ethylenediamine-N,N′-diacetic acid, ≥98%
C6H12N2O4