EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Simultaneous purification and site-specific modification of pyrroline-carboxy-lysine proteins.
PMID 22223621
Abstract
Sticky residue: Pyrroline-carboxy-lysine (Pcl) can be readily incorporated into proteins expressed in E. coli and mammalian cells by using the pyrrolysyl tRNA/tRNA synthetase pair. Pcl can be used as a single amino acid purification tag and can be site-specifically modified with functional probes during the elution process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9628
2-Aminobenzaldehyde, ≥98%
C7H7NO