EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Evidence by chemical modification that tryptophan-104 of the cysteine-proteinase inhibitor chicken cystatin is located in or near the proteinase-binding site.
PMID 2222419
Abstract
The single tryptophan residue Trp-104 of chicken cystatin was modified with a 2-hydroxy-5-nitrobenzyl group. The change of the absorption spectrum of this group on binding of the modified cystatin to papain indicated a decreased environmental polarity of the probe. The modified inhibitor had about a 10(5)-fold lower affinity for papain than had intact cystatin, this being due to a higher dissociation rate constant. These results show that Trp-104 of cystatin is located in or near the proteinase-binding site of the inhibitor, in agreement with a model proposed from computer docking Experiments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6388
Dimethyl(2-hydroxy-5-nitrobenzyl)sulfonium bromide, ≥95%
C9H12BrNO3S