EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Update on the development of antagonists of chemoattractant receptor-homologous molecule expressed on Th2 cells (CRTH2). From lead optimization to clinical proof-of-concept in asthma and allergic rhinitis.
PMID 22224640
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0531
Ramatroban, ≥98% (HPLC), solid
C21H21FN2O4S