EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Engineering of a tyrosol-producing pathway, utilizing simple sugar and the central metabolic tyrosine, in Escherichia coli.
PMID 22225426
Abstract
Metabolic engineering was applied to the development of Escherichia coli capable of synthesizing tyrosol (2-(4-hydroxyphenyl)ethanol), an attractive phenolic compound with great industrial value, from glucose, a renewable carbon source. In this strain, tyrosine, which was supplied not only from the culture medium but also from the central metabolism, was converted into tyrosol via three steps: decarboxylation, amine oxidation, and reduction. The engineered strain synthesized both tyrosol and 4-hydroxyphenylacetate (4HPA), but disruption of the endogenous phenylacetaldehyde dehydrogenase gene shut off 4HPA production and improved the production of tyrosol as a sole product. The engineered mutant strain was capable of producing 0.5 mM tyrosol from 1% (w/v) glucose during a 48 h shake flask cultivation.
Related Materials