EMAIL THIS PAGE TO A FRIEND
Methods in cell biology
Culture and manipulation of embryonic cells.
PMID 22226523
Abstract
The direct manipulation of embryonic cells is an important tool for addressing key questions in cell and developmental biology. C. elegans is relatively unique among genetic model systems in being amenable to manipulation of embryonic cells. Embryonic cell manipulation has allowed the identification of cell interactions by direct means, and it has been an important technique for dissecting mechanisms by which cell fates are specified, cell divisions are oriented, and morphogenesis is accomplished. Here, we present detailed methods for isolating, manipulating and culturing embryonic cells of C. elegans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9523
Sodium cholesteryl sulfate
C27H45NaO4S