EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Impact of herbicide Ametryn on microbial communities in mixed liquor of a membrane bioreactor (MBR).
PMID 22230781
Abstract
Ametryn, which is a second generation herbicide, was introduced to a lab-scale MBR at a concentration of 1mg/L and a 20-40% removal was observed at HRT ranging from 7.8 to 15.6h for an average influent Ametryn concentration of 0.8 mg/L. Components of EPS (protein and carbohydrates) increased in the bioreactor and the observed biomass production reduced after the addition of Ametryn. In a batch study, GAC was added to MBR mixed liquor and removal of Ametryn via biodegradation and adsorption were measured. Five common bacterial colony types (Gram negative and positive bacilli and Gram negative cocci) were identified and three of these were resistant to Ametryn up to 5mg/L. GAC was found to be a very effective Ametryn adsorption medium and in some occasions Ametryn may have acted as a nutrient source for bacteria.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45321
Ametryn, PESTANAL®, analytical standard
C9H17N5S
PS383
Ametryn, analytical standard
C9H17N5S