EMAIL THIS PAGE TO A FRIEND
Acta pharmacologica Sinica
Novel agents inhibit human leukemic cells.
PMID 22231394
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6377
1-Hydroxypyridine-2-thione zinc salt, ≥95%
C10H8N2O2S2Zn