EMAIL THIS PAGE TO A FRIEND
Journal of pediatric gastroenterology and nutrition
Visceral hypersensitivity in IBS: from bench to bedside.
PMID 22235465
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K2628 Ketotifen fumarate salt
C19H19NOS · C4H4O4