EMAIL THIS PAGE TO A FRIEND
Contraception
Pelvic surgery and hospitalization among Chilean women after nonsurgical sterilization with quinacrine pellets between 1977 and 1989.
PMID 22240176
Abstract
Concern about quinacrine lingers because of its carcinogenic effects in rats. We describe results of long-term follow-up of women who underwent quinacrine pellet sterilization in Chile between 1977 and 1989 (N=1492). We interviewed the women or relatives in five rounds of data collection between 1991-1993 and 2006-2007, and reviewed hospital records. Median follow-up was 18.5 years; total person-time was 23,894 woman-years. This analysis focuses on pelvic and abdominal surgeries and conditions. We used survival analysis to estimate the 15-year cumulative probability of hysterectomy, other pelvic surgical procedures and relevant adverse events. Uterine fibroids were by far the most common gynecologic condition, reported by 11% of the cohort. Surgical procedures were recorded for 15% of the cohort; hysterectomy was the most frequent procedure (10%), followed by salpingectomy (2%). The 15-year probability of any pelvic or abdominal procedure was 14.7 per 100 women (95% confidence interval 12.4-16.9). The probability of hysterectomy was 9.3 per 100 women (95% confidence interval 7.4-11.1). Number of quinacrine insertions had little impact on the probabilities. During long-term follow-up of women who received quinacrine pellets for nonsurgical sterilization, the incidence of noncancer adverse outcomes was not unusually high, and no alarming patterns emerged.