EMAIL THIS PAGE TO A FRIEND
International journal of cardiology
HMGB1: a potential therapeutic target for myocardial ischemia and reperfusion injury.
PMID 22240761
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W245712
Ethyl pyruvate, natural, FG
C5H8O3
E47808
Ethyl pyruvate, 98%
C5H8O3