EMAIL THIS PAGE TO A FRIEND
Anti-cancer drugs
Ampelopsin sodium exhibits antitumor effects against bladder carcinoma in orthotopic xenograft models.
PMID 22241170
Abstract
The aim of this study was to establish xenograft models of tumor in mice bladder and evaluate the antitumor efficacy of ampelopsin sodium (Amp-Na). A total of 2×10 human bladder carcinoma EJ cells and murine sarcoma 180 cells were instilled into the bladder of BALB/c nu/nu mice and Swiss mice after preconditioning to establish the tumor model. Mice bearing orthotopic tumors were treated with Amp-Na by intravenous, intraperitoneal, or intravesical instillation. In addition, the pharmacokinetics property of Amp-Na was investigated in normal BALB/c mice. Our results showed that Amp-Na was excreted mainly through the urine, where it existed at a high concentration. Amp-Na significantly inhibited the proliferation of EJ and sarcoma 180 cells both in vivo and in vitro and this can be at least partially attributed to the cell cycle arrest induced by Amp-Na. This study suggests that the use of Amp-Na is an attractive chemotherapeutic modality for bladder cancer patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0295
Dihydromyricetin, ≥98% (HPLC)
C15H12O8