EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[Iridoid glycosides from buds of Jasminum officinale L. var. grandiflorum].
PMID 22242454
Abstract
The study on the buds of Jasminum officinale L. var. grandiflorum was carried out to look for anti-HBV constituents. The isolation and purification were performed by HPLC and chromatography on silica gel, polyamide and Sephadex LH-20 column. The structures were elucidated on the basis of physicochemical properties and spectral analysis. Six iridoid glycosides were identified as jasgranoside B (1), 6-O-methy-catalpol (2), deacetyl asperulosidic acid (3), aucubin (4), 8-dehydroxy shanzhiside (5), and loganin (6). Jasgranoside B (1) is a new compound. Compounds 2-6 were isolated from Jasminum officinale L. var. grandiflorum for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36483
Loganin, analytical standard
C17H26O10