EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Phospha-Michael additions to activated internal alkenes: steric and electronic effects.
PMID 22242695
Abstract
The addition of P(O)-H bonds to internal alkenes has been accomplished under solvent-free conditions without the addition of a catalyst or radical initiator. Using a prototypical secondary phosphine oxide, a range of substrates including cinnamates, crotonates, coumarins, sulfones, and chalcones were successfully functionalized. Highly activated acceptors such as isopropylidenemalononitrile and ethyl 2-cyano-3-methyl-2-butenoate underwent the phospha-Michael reaction upon simple trituration of the reagents at room temperature, whereas less activated substrates such as ethyl cinnamate and methyl crotonate required heating (>150 °C) in a microwave reactor to achieve significant consumption of the starting alkenes. For the latter alkenes, a competing reaction involving disproportionation of the ditolylphosphine oxide into ditolylphosphinic acid and ditolylphosphine was observed at the high temperatures needed to promote the addition reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
310581
Ethyl 2-cyano-3-methyl-2-butenoate, 97%
C8H11NO2