EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering
Studies on the removal of Lissamine Green B from clays and soil in comparison with contemporary approaches.
PMID 22242878
Abstract
A combination of Fenton's reagent with electrochemistry has been demonstrated to be a efficient method for removing a dye (Lissamine Green B) from clays (kaolin, montmorillonite) and soil. The two-step approach described here involved quantitative extraction with hot water, followed by quantitative removal of the Lissamine Green dye by column chromatography using Octolig®. The advantage of this procedure is success without the need for Fenton reagents or electricity. A disadvantage is the process would not work with polycyclic hydrocarbons such as phenanthrene, though the electro-Fenton degradation does.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06737
Green S, analytical standard
C27H25N2NaO7S2
199583
Lissamine Green B, Dye content 60 %
C27H25N2NaO7S2