EMAIL THIS PAGE TO A FRIEND
Journal of food science
Changes in flavor precursors, pungency, and sugar content in short-day onion bulbs during 5-month storage at various temperatures or in controlled atmosphere.
PMID 22242977
Abstract
Short-day onion bulbs (cv. TG 1015Y) were stored in 1% O(2,) 99% N(2) air at 5 °C (controlled atmosphere [CA]), or in ambient air at 5, 24, or 30 °C, for 5 mo. Changes in flavor precursors, pungency, and sugar content were investigated. After 5 mo of storage, 1-propenyl-L-cysteine sulfoxide concentrations increased continuously at 5 °C, gradually decreased in CA, slightly increased for 3 mo, and returned to initial levels at 24 °C and decreased below initial levels at 34 °C. Methyl-L-cysteine sulfoxide concentrations remained unchanged in all storage conditions. Onion pungency levels significantly increased at 5 °C, and decreased at 30 °C. Storage in CA and at 24 °C resulted in some fluctuations in pungency but the levels remained similar to initial levels. The calculated pyruvic acid concentrations were approximately 1.0 μmole/mL higher than the measured concentrations, and showed an increase at 5 °C and a slight reduction at 30 °C, consistent with the pungency results. Storage at 5 °C and in CA resulted in slight increases in fructose and glucose concentrations for 3 to 4 mo with subsequent rapid decreases, while sucrose concentrations remained unchanged. However, at 24 and 30 °C, fructose and glucose concentrations continuously decreased, accompanied by a continuous increase in sucrose concentrations. Storage in CA maintained the quality of onions best, as evidenced by the smallest changes in flavor precursors, pungency, and sugar concentrations, while storage at 5 °C resulted in increased pungency. Storage at 24 and 30 °C was tested for the purpose of comparison only; these temperatures are not recommended for commercial storage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
410209
L-Cysteine methyl ester hydrochloride, 98%
C4H9NO2S · HCl