EMAIL THIS PAGE TO A FRIEND
Bioconjugate chemistry
Novel cyanine dyes with vinylsulfone group for labeling biomolecules.
PMID 22243689
Abstract
Novel vinylsulfone cyanine dyes (em. 550-850 nm) were designed and synthesized for fluorescent labeling of biomolecules via 1,2-addition reaction in aqueous conditions. Due to the virtue of chemical structures of both fluorophore and reactive group, these dyes could be significantly stable and reactive in various aqueous/organic conditions. A wide variety of pH, temperature, buffer concentration, and protein were tested for the optimal labeling condition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V3700
Divinyl sulfone, contains 400-600 hydroquinone as inhibitor, ≥96%
C4H6O2S