EMAIL THIS PAGE TO A FRIEND
Fitoterapia
Synergistic antibiotic activity of volatile compounds from the essential oil of Lippia sidoides and thymol.
PMID 22245085
Abstract
Lippia sidoides Cham. (Verbenaceae) is used in the folk medicine as topical antiseptic in skin and mucous membranes and its therapeutic effect is attributed to the thymol presence. The objective of this work was to verify the chemical composition and antibiotic modifying activity of the essential oil extracted from the leaves of L. sidoides and its major component thymol. The essential oil was obtained by hydrodistillation and analyzed by GC/MS. The synergistic activity was evaluated using gaseous contact method. The essential oil was obtained (yield of 1.06%) and the GC/MS analysis identified the main constituents: thymol (84.9%) and p-cymene (5.33%). The antibiotic modifying activity was verified using the minimal inhibitory dose method and gaseous contact. It verified the interference of essential oil and thymol against all tested aminoglycosides. There were no statistical differences between the activity of the essential oil and thymol against Pseudomonas aeruginosa, indicating this to be the responsible composition for such activity. However, the oil was shown more effective when compared to the thymol against Staphylococcus aureus. The essential oil of L. sidoides and its major component thymol influence the activity of aminoglycosides and may be used as adjuvant in antibiotic therapy against respiratory tract bacterial pathogens.