EMAIL THIS PAGE TO A FRIEND
Plant physiology and biochemistry : PPB
Salicylic acid is a modulator of catalase isozymes in chickpea plants infected with Fusarium oxysporum f. sp. ciceri.
PMID 22245913
Abstract
The relationship between salicylic acid level catalases isoforms chickpea cv. ICCV-10 infected with Fusarium oxysporum f. sp. ciceri was investigated. Pathogen-treated chickpea plants showed high levels of SA compared with the control. Two isoforms of catalases in shoot extract (CAT-IS and CAT-IIS) and single isoform in root extract (CAT-R) were detected in chickpea. CAT-IS and CAT-R activities were inhibited in respective extracts treated with pathogen whereas, CAT-IIS activity was not inhibited. These isoforms were purified and their kinetic properties studied in the presence or absence of SA. The molecular mass determined by SDS-PAGE of CAT-IS, CAT-IIS and CAT-R was found to be 97, 40 and 66 kDa respectively. Kinetic studies indicated that Km and V(max) of CAT-IS were 0.2 mM and 300 U/mg, 0.53 mM and 180 U/mg for CAT-IIS and 0.25 mM and 280 U/mg for CAT-R, respectively. CAT-IS and CAT-R were found to be more sensitive to SA and 50% of their activities were inhibited at 6 and 4 μM respectively, whereas CAT-IIS was insensitive to SA up to 100 μM. Quenching of the intrinsic tryptophan fluorescence of purified catalases were used to quantitate SA binding; the estimated K(d) value for CAT-IS, CAT-IIS and CAT-R found to be 2.3 μM, 3.1 mM and 2.8 μM respectively. SA is a modulator of catalase isozymes activity, supports its role in establishment of SAR in chickpea plants infected with the pathogen.
Related Materials