EMAIL THIS PAGE TO A FRIEND
Journal of bioenergetics and biomembranes
Prerequisites for ubiquinone analogs to prevent mitochondrial permeability transition-induced cell death.
PMID 22246424
Abstract
The permeability transition pore (PTP) is a mitochondrial inner membrane channel involved in cell death. The inhibition of PTP opening has been proved to be an effective strategy to prevent cell death induced by oxidative stress. Several ubiquinone analogs are known to powerfully inhibit PTP opening with an effect depending on the studied cell line. Here, we have studied the effects of ubiquinone 0 (Ub(0)), ubiquinone 5 (Ub(5)) and ubiquinone 10 (Ub(10)) on PTP regulation, H(2)O(2) production and cell viability in U937 cells. We found that Ub(0) induced both PTP opening and H(2)O(2) production. Ub(5) did not regulate PTP opening yet induced H(2)O(2) production. Ub(10) potently inhibited PTP opening yet induced H(2)O(2) production. Both Ub(0) and Ub(5) induced cell death, whereas Ub(10) was not toxic. Moreover, Ub(10) prevented tert-butyl hydroperoxide-induced PTP opening and subsequent cell death. We conclude that PTP-inhibitor ubiquinone analogs are able to prevent PTP opening-induced cell death only if they are not toxic per se, which is the case when they have no or low pro-oxidant activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9150
2,3-Dimethoxy-5-methyl-p-benzoquinone, apoptosis inducer
C9H10O4
C7956 Coenzyme Q1, ≥95%
C14H18O4