EMAIL THIS PAGE TO A FRIEND
American journal of respiratory and critical care medicine
Septic pulmonary embolism in a patient with defibrillator lead endocarditis.
PMID 22246711
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32071
Nafcillin sodium salt, VETRANAL®, analytical standard
C21H21N2NaO5S