EMAIL THIS PAGE TO A FRIEND
European journal of neurology
Is there a future for neuroprotective agents in acute ischaemic stroke?
PMID 22248300
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
01518
Ginsenoside Rd, analytical standard
C48H82O18