EMAIL THIS PAGE TO A FRIEND
Journal of peptide science : an official publication of the European Peptide Society
Stereoselective synthesis of fully protected (2S,4S,6S)-2-amino-6-hydroxy-4-methyl-8-oxodecanoic acid (AHMOD).
PMID 22249935
Abstract
The stereocontrolled synthesis of fully protected (2S,4S,6S)-2-amino-6-hydroxy-4-methyl-8-oxodecanoic acid was accomplished using a glutamate derivative as starting material. The key steps of this stereochemical synthetic pathway involved an Evans asymmetric alkylation, a Sharpless asymmetric epoxidation, and a Grignard reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4151
Sodium decanoate, ≥98%
C10H19NaO2