EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
New 2, 6-modified Bodipy sensitizers for dye-sensitized solar cells.
PMID 22250129
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
647292
4-(Diphenylamino)phenylboronic acid, ≥95%
C18H16BNO2