EMAIL THIS PAGE TO A FRIEND
Nihon Naika Gakkai zasshi. The Journal of the Japanese Society of Internal Medicine
[Case report: a case of anterocollis ameliorated by trihexyphenidyl].
PMID 22250425
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000108
Trihexyphenidyl hydrochloride, European Pharmacopoeia (EP) Reference Standard
C20H31NO · HCl