EMAIL THIS PAGE TO A FRIEND
Natural product research
Comparative study on the antioxidant activities of the different extracts and the composition of the oil extracted by n-hexane from Iranian Vitex pseudo-negundo.
PMID 22250599
Abstract
Antioxidant activities of different extracts obtained from the aerial parts of Vitex pseudo-negundo from Kashan, central Iran, were evaluated for the first time in this study using β-carotene/linoleic acid and scavenging of free-radical (DPPH) assays. Water extract showed the highest activity in both assays. GC-MS analysis of the oil extracted by n-hexane revealed 46 compounds with trans-β-farnesene being the main component. Several new compounds, not reported in the previous literature, were identified in the essential oil of this chemo-type.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73492
trans-β-Farnesene, analytical standard
C15H24