EMAIL THIS PAGE TO A FRIEND
Animal science journal = Nihon chikusan Gakkaiho
Effects of a combination of hinokitiol (β-thujaplicin) and an organic acid mixture on ruminal fermentation in heifers fed a high-grain diet.
PMID 22250737
Abstract
This study evaluated the effects of hinokitiol (a natural antibacterial compound extracted from Thujopsis dolabrata var. hondai) and an organic acid mixture (citrate content 50%) on ruminal fermentation. Antibacterial properties were examined by measuring minimal inhibitory concentration. Hinokitiol at 1.56µg/mL or an organic acid mixture at 1600µg/mL inhibited Streptococcus bovis growth. The combination of 0.78µg/mL hinokitiol and 200µg/mL of an organic acid mixture also inhibited S. bovis growth. Both hinokitiol and the hinokitiol and an organic acid mixture combination showed strong antibacterial properties on Gram-positive bacteria such as S. bovis, but relatively weak antibacterial activities on Gram-negative bacteria such as Megasphaera elsdenii. Three ruminally cannulated heifers were fed a bloat-producing diet containing barley, pelleted alfalfa meal, soybean meal and salt without long-cut roughage to investigate the ruminal characteristics in vivo. Feeding to heifers a bloat-producing diet containing 7.8mg/kg hinokitiol and 0.2% of an organic acid mixture significantly decreased the increase in stable ingesta volume. Hinokitiol or an organic acid mixture did not affect ruminal volatile fatty acids, protozoa and bacteria. These results suggest that a combination of hinokitiol and an organic acid mixture might reduce frothy bloat in cattle fed high-grain diets.