EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Therapeutic potential of selenium and tellurium compounds: opportunities yet unrealised.
PMID 22252404
Abstract
Despite being disparaged for their malodorous and toxic demeanour, compounds of selenium, a bio-essential element, and tellurium, offer possibilities as therapeutic agents. Herein, their potential use as drugs, for example, as anti-viral, anti-microbial, anti-inflammatory agents, etc., will be surveyed along with a summary of the established biological functions of selenium. The natural biological functions of tellurium remain to be discovered.