EMAIL THIS PAGE TO A FRIEND
Journal of anesthesia
Vecuronium was safely used in a patient with Engelmann's disease without muscle weakness.
PMID 22252646
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76904
Vecuronium bromide
C34H57BrN2O4
Y0000561
Vecuronium bromide, European Pharmacopoeia (EP) Reference Standard
C34H57BrN2O4