EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Determination of hyperoside and isoquercitrin in rat plasma by membrane-protected micro-solid-phase extraction with high-performance liquid chromatography.
PMID 22258808
Abstract
A novel method, micro-solid-phase extraction based on membrane-protected molecularly imprinted polymer, was developed to extract hyperoside and isoquercitrin in rat plasma. Synthesized hyperoside MIPs were packed in a porous polyether sulfone membrane envelope to perform extraction. The parameters sorbent materials, membrane types, extraction time and desorption conditions were optimized for micro-solid-phase extraction. Under the optimal conditions, correlation coefficients, 0.998 and 0.999, were obtained for hyperoside and isoquercitrin, respectively, with the linear range between 1 and 120 μg/mL. The absolute extraction recoveries from 84.5 to 89.3% were found. The method detection limits of hyperoside and isoquercitrin were 0.24 and 0.22 μg/mL, respectively. Compared with traditional methods, solid-phase extraction, liquid-liquid extraction and protein precipitation, the developed method was simple, highly efficient for extraction, environmentally friendly, and particularly suitable for complex biological samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00180585
Hyperoside, primary reference standard
C21H20O12
83388
Quercetin 3-D-galactoside, ≥97.0% (HPLC)
C21H20O12