EMAIL THIS PAGE TO A FRIEND
Talanta
Pressurised hot water extraction followed by headspace solid-phase microextraction and gas chromatography-tandem mass spectrometry for the determination of N-nitrosamines in sewage sludge.
PMID 22265500
Abstract
A method for the quantitative determination of the nine EPA N-nitrosamines in sewage sludge was developed by using pressurised hot water extraction (PHWE) followed by headspace solid-phase microextraction (HS-SPME) and gas chromatography coupled to chemical ionization ion trap tandem mass spectrometry (GC-CI-MS-MS). The pressurised hot water extraction was optimized using a central composite design with regard to operational parameters such as temperature, extraction time and pH of water as extracting solvent. The optimum conditions were: water at pH 7.5 as extracting solvent, temperature of 125°C and extraction time of 5 min. The sewage sludge extract was automatically analyzed by HS-SPME using a divinylbenzene/carboxen/polydimethylsiloxane (DVB/CAR/PDMS) fiber and GC-CI-MS-MS. The limits of detection of all compounds were lower than 0.15 μg/kg of dry weight (d.w.) of sewage sludge. The repeatability and reproducibility between days (10μg/kg d.w.) expressed as relative standard deviation were lower than 16 and 19%, respectively. The method was applied to determine the N-nitrosamines in sewage sludge from urban and industrial sewage treatment plants (STPs) and from a potable water treatment plant. Some N-nitrosamines were determined in the samples and N-nitrosodiethylamine (NDEA) and N-nitrosodi-n-butylamine (NDBA) showed the highest values (371 and 305 μg/kg (d.w.), respectively) in sewage from industrial STPs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
57328-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
57348-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with manual holder/autosampler
57329-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
57299-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder/autosampler, fiber L 2 cm
57298-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
57912-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 1 cm
57914-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm
FFA57298-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, for use with multi fiber exchanger, Stableflex
FFA57329-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with multi fiber exchanger