EMAIL THIS PAGE TO A FRIEND
Steroids
A simple and convenient synthesis of 2-methoxyestradiol from estrone.
PMID 22266333
Abstract
A simple and straightforward synthesis of 2-methoxyestradiol have been achieved in nine synthetic steps with 21% of overall yield. Being a convenient process, it can be upscaled to industrial process.