EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Palladium-catalyzed cross-coupling reactions of organogold(I) phosphanes with allylic electrophiles.
PMID 22266785
Abstract
Aryl and alkenylgold(I) phosphanes react regioselectively with allylic electrophiles such as cinnamyl and geranyl halides (bromide, chloride and acetates) under palladium catalysis in THF at 80 °C to afford the α-substitution product with moderate to high yields. When the reaction is performed with a chiral enantiopure secondary acetate, the α-substituted cross-coupling product is obtained with complete inversion of the stereochemistry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
187186
Cinnamyl chloride, 95%
C9H9Cl
329118
Geranyl bromide, 95%
C10H17Br
302791
Geranyl chloride, 95%
C10H17Cl