EMAIL THIS PAGE TO A FRIEND
Archiv der Pharmazie
In vivo antihyperlipidemic activity of a new series of N-(benzoylphenyl) and N-(acetylphenyl)-1-benzofuran-2-carboxamides in rats.
PMID 22266816
Abstract
A new series of N-(benzoylphenyl) and N-(acetylphenyl)-1-benzofuran-2-carboxamides (3a-3d and 4a'-4c') were synthesized. Compounds (3a, 3b, and 4a'-4c') were tested in vivo using Triton-WR-1339-induced hyperlipidemic rats as an experimental model for their hypolipidemic activity. The tested animals were divided into eight groups: control, hyperlipidemic, 3a, 3b, 4a', 4b', 4c', and bezafibrate. At a dose of 15 mg/kg, the elevated plasma triglyceride (TG) levels were significantly reduced in compounds 3b (p <0.0001) and 4c' (p <0.05) after 12 and 24 h compared to the normal control group. Furthermore, high-density lipoprotein-cholesterol levels were remarkably increased in compounds 3b (p <0.001) and 4c' (p <0.05). Meanwhile, compound 4b' slightly reduced the TG levels after 12 and 24 h. The present study demonstrated new properties of the novel series of benzofuran-2-carboxamides 3b and 4c' as potent lipid-lowering agents. It is, therefore, reasonable to assume that compounds 3b and 4c' may have a promising potential in the treatment of hyperlipidemia and coronary heart diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8761
Tyloxapol, nonionic surfactant
T0307
Tyloxapol, BioXtra