EMAIL THIS PAGE TO A FRIEND
European journal of ophthalmology
Endogenous endophthalmitis with an unusual infective agent: Actinomyces neuii.
PMID 22267451
Abstract
To report an unusual case of a patient with endogenous endophthalmitis caused by Actinomyces neuii. A 69-year-old woman in an immunosuppressed state and who had a previous history of periappendicular abscess presented with bilateral red painful eyes. The diagnosis was confirmed by culture and pan-bacterial polymerase chain reaction drawn from anterior chamber sample. On admission, the patient underwent an intravitreal injection of vancomycin combined with ceftazidime. Following a 3-week treatment of intravenous penicillin and topical sulfacetamide sodium, the patient recovered fully. Actinomyces neuii can cause endogenous endophthalmitis. Intravenous penicillin G is an effective treatment leading to favorable prognosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46770
Sulfacetamide, VETRANAL, analytical standard
C8H10N2O3S
S8647
Sulfacetamide sodium salt monohydrate
C8H9N2NaO3S · H2O