EMAIL THIS PAGE TO A FRIEND
Pest management science
A non-destructive method for testing two components of the behaviour of soil-applied agricultural chemicals over a long period.
PMID 22271588
Abstract
Owing to the complexity of soil composition, accurate predictions of both apoplastic systemicity of lipophilic xenobiotics and their leaching from the soil are made difficult. Therefore, a non-destructive method to assess directly these two components of the spatial behaviour of soil-applied phytochemicals is needed. The plant selected was a dwarf tomato, which can exude an abundant apoplastic fluid through large stomata for several months. The feasibility and reliability of the method were assayed using three triazoles exhibiting different log D values. HPLC-MS analyses indicated that triadimenol (log D = 2.97) was clearly the most mobile compound within the apoplast, especially its diastereoisomer A. Propiconazole (log D = 3.65) and penconazole (log D = 4.64) exhibited a similar low systemicity. The data remained the same when the three fungicides were applied together on the soil. Long time-course studies (1.5 months) of penconazole behaviour indicated that, in contrast to leaching, which decrease sharply, root-to-shoot translocation remained almost unchanged during the whole experiment, in spite of the high lipophilicity of this fungicide. This method must contribute to a better knowledge of the behaviour of commercial soil-applied phytochemicals. It can also be used to screen new xenobiotics within strategies to satisfy environmental requirements.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36189
Penconazol, PESTANAL®, analytical standard
C13H15Cl2N3