EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Development of a novel monolith frit-based solid-phase microextraction method for determination of hexanal and heptanal in human serum samples.
PMID 22271665
Abstract
In this paper, a polypropylene frit with porous network structure and high area-to-thickness ratio (4.8 mm diameter, 1.6 mm thickness, 20 mm pore size) was utilized as a mould of monolith. Poly(methacrylic acid-ethlyene glycol dimethacrylate) (MAA-EGDMA) monolith was in situ synthesized in the micro-channel of frit by photopolymerization. A monolith frit-based solid-phase microextraction method (SPME) was developed for the determination of hexanal and heptanal in serum samples by combining with high-performance liquid chromatography. 2,4-Dinitrophenylhydrazine (DNPH) as the derivatizing reagent was absorbed on a monolith frit, then its derivatization reaction with aldehydes and the absorption of formed hydrazones on the monolith disk occurred simultaneously. The condition parameters for polymerization, derivatization and extraction were optimized systematically. Under the optimum conditions, rigid structure, low back-pressure and high column capacity were achieved for the monolith frit. The limits of detection for hexanal and heptanal were 1.86 and 1.38 nmol/L, respectively. The inter- and intra-day relative standard deviations were less than 7.7% (n = 6). This method was applied successfully to aldehydes analysis in human serum samples. The method possesses advantages such as simplicity, efficiency, low cost and good biocompatibility. It provides an alternative approach for quantification of aldehydes in complex biological samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H2120
Heptaldehyde, 95%
C7H14O
W254002
Heptanal, ≥95%, FCC, FG
C7H14O