EMAIL THIS PAGE TO A FRIEND
Current Alzheimer research
In vivo uptake of β-amyloid by non-plaque associated microglia.
PMID 22272621
Abstract
The role of microglia in β-amyloid (Aβ) deposition or clearance in the Alzheimer's disease (AD) brain remains unclear. Previous in vivo studies have focused primarily on the association of microglia with Aβ-positive parenchymal plaques, but have given little consideration to the possible interaction between Aβ and non-plaque associated microglia. Further, it is not known if microglia play a direct role in mediating Aβ uptake following anti-aggregant treatment. We report here the identification of Aβ-positive processes throughout the cortex and hippocampus of TgCRND8 mice expressing the human Swedish (KM670/671NL) and Indiana (V717F) amyloid precursor protein mutations, which localized to ionized calcium binding protein-1-positive resident microglia that were not associated with extracellular plaques. Oral administration of 1-deoxy-1-fluoro-scyllo-inositol, a scyllo-inositol analogue, to TgCRND8 mice improved spatial memory impairments and suppressed amyloid pathology in a dose-dependent manner. Further, treatment with 1-deoxy-1- fluoro-scyllo-inositol significantly increased hippocampal intra-microglial Aβ levels without stimulating microglial proliferation or peripheral macrophage recruitment. These results reveal a novel, beneficial role for non-plaque associated microglia in the regulation of cerebral Aβ levels in a mouse model of AD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I8132
scyllo-Inositol, ≥98%
C6H12O6