EMAIL THIS PAGE TO A FRIEND
Chemosphere
Whole blood is the sample matrix of choice for monitoring systemic triclocarban levels.
PMID 22273184
Abstract
The antibacterial triclocarban (TCC) concentrates in the cellular fraction of blood. Consequently, plasma levels are at least two-fold lower than the TCC amount present in blood. Utilizing whole blood sampling, a low but significant absorption of TCC from soap during showering is demonstrated for a small group of human subjects.