EMAIL THIS PAGE TO A FRIEND
Organic letters
Copper-catalyzed [5+1] annulation of 2-ethynylanilines with an N,O-acetal leading to construction of quinoline derivatives.
PMID 22276849
Abstract
A novel copper-catalyzed [5 + 1] annulation of 2-ethynylanilines with an N,O-acetal, which functioned as a C1 part, leading to the preparation of quinoline derivatives with an ester substituent on the 2-position is described. A combination of CuBr(2) and trifluoroacetic acid (TFA) promoting [5 + 1] annulation of the 2-ethynylaniline with ethyl glyoxylate is also demonstrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
260150
Glyoxylic acid solution, 50 wt. % in H2O
C2H2O3