EMAIL THIS PAGE TO A FRIEND
Supportive care in cancer : official journal of the Multinational Association of Supportive Care in Cancer
Savene® (dexrazoxane) use in clinical practice.
PMID 22278308
Abstract
Anthracycline extravasation (ACEV) is a rare but potentially devastating event which can result in severe injuries including ulceration and necrosis, slow-healing lesions, serious joint damage and permanent disfigurement. It can delay further scheduled chemotherapy and affect cancer treatment outcome. Savene® (dexrazoxane) is the only approved antidote for ACEV in Europe (Totect® in the USA) and is administered by intravenous infusion. Its efficacy has been demonstrated in clinical trials with biopsy-verified ACEV with a 98% success rate (no need for surgical debridement) allowing for immediate continuation of chemotherapy in 71% of patients. Adverse events, mainly haematological toxicity, were rapidly reversible. The objective of the study was to assess, in clinical practice, the efficacy and safety profile of Savene® for ACEV in different Belgian hospitals. A survey of Savene® use was conducted in Belgium from 2007 to 2010 by using a questionnaire sent to 44 hospitals. Data were obtained for 41 cancer patients, 68% (28/41) had ACEV from central venous catheters. Surgical debridement due to ACEV could be avoided in 26 out of 28 extravasations from a central venous access and in 95% (39/41) of the total population treated with Savene®. Planned chemotherapy was maintained in 73% (30/41) of patients. Eight adverse events were reported in four patients treated with Savene®, six events were assessed to be of common toxicity criteria grades 1-2 (nausea, leucopenia and arm pain) and two events (neutropenia and pancytopenia) were assessed to be grade 3. These data are comparable with the data from previous clinical trials and confirm the efficacy and safety profile of Savene® in clinical practice for the treatment of anthracycline extravasation, including extravasations from central venous catheters.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1446
Dexrazoxane, ≥95% (HPLC)
C11H16N4O4
R8657
Razoxane, >98% (HPLC)
C11H16N4O4