EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Enhancement of ferroelectric polarization stability by interface engineering.
PMID 22278910
Abstract
By using theoretical predictions based on first-principle calculations, we explore an interface engineering approach to stabilize polarization states in ferroelectric heterostructures with a thickness of just several nanometers.