EMAIL THIS PAGE TO A FRIEND
Sleep medicine
Breakthrough symptoms during the daytime in patients with restless legs syndrome (Willis-Ekbom disease).
PMID 22281003
Abstract
It is often assumed that most patients with restless legs syndrome (RLS) only experience symptoms at night. However, previous studies have estimated the prevalence of daytime symptoms to be 10-60%. This study sought to investigate the prevalence and pattern of daytime symptoms in patients with moderate-to-severe RLS. Observational, cross-sectional investigation. A self-administered questionnaire was sent out, on a random basis, to 310 patients with RLS by the Spanish RLS patient support group. Only individuals with a confirmed diagnosis of RLS were included in the final survey. In total, 224 individuals were included in the survey (response rate 72%). Over 55% of patients reported daytime crises on most (>3) days of the week, and 41% suffered daytime symptoms on a daily basis. These breakthrough crises were characterized by unexpected and sudden symptoms and were frequently precipitated by a reduction in daytime activity. The mean severity of these crises on a visual analogue scale (range 0-10) was 6.8 (standard deviation 2.1), and they had a major impact on quality of life. The prevalence of breakthrough crises was related to duration of illness but not to duration of treatment. This study suggests that breakthrough crises are common in moderate-to-severe RLS and have a negative effect on quality of life. More studies are needed to investigate whether breakthrough crises reflect disease progression or, at least for those patients undergoing dopaminergic treatment, whether they represent an early indication of RLS augmentation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R2530
Ropinirole hydrochloride, powder, ≥98% (HPLC)
C16H24N2O · HCl
R9281
Rotigotine hydrochloride, ≥98% (HPLC)
C19H25NOS·HCl