EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
Homozygous familial hypercholesterolaemia.
PMID 22285056
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4650 Cholestyramine resin