EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Chemical generation of o-quinone monoimines for the rapid construction of 1,4-benzoxazine derivatives.
PMID 22286300
Abstract
Highly reactive o-benzoquinone monoimines were chemically generated and successfully trapped with electron-rich olefins that led to the synthesis of hitherto unknown 1,4-benzoxazine derivatives. This unprecedented transformation was achieved by the oxidation of o-aminophenols bearing appropriate functionality on the arene residue with less expensive hypervalent iodine reagent in the presence of vinylic ethers or thioethers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09120
2-Aminophenol, purum, ≥98.0% (HPLC)
C6H7NO
A71301
2-Aminophenol, 99%
C6H7NO