EMAIL THIS PAGE TO A FRIEND
Plastic and reconstructive surgery
Antibacterial analysis of surgical adhesives.
PMID 22286489
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0878 Gum mastic