EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Cloning and sequencing of inulinase and β-fructofuranosidase genes of a deep-sea Microbulbifer species and properties of recombinant enzymes.
PMID 22286980
Abstract
An inulinase-producing Microbulbifer sp. strain, JAM-3301, was isolated from a deep-sea sediment. An inulin operon that contained three open reading frames was cloned and sequenced. Two of the three genes were expressed. One product was an endo-inulinase, and the other was a β-fructofuranosidase. Both enzymes worked together to effectively degrade inulin.
Related Materials